Stampa…
Dosing spoons, white
Fornitore: BURKLE
5378-4014 5378-4015 5378-4017 5378-4019 5378-4006 5378-4007 5378-4011 5378-4012 5378-4013 5378-4010 5378-4016 5378-4018 5378-4004 5378-4008 5378-4009 5378-4005
300-0372EA 53.5 EUR
300-0372 300-0373 300-0375 300-0377 300-0364 300-0365 300-0369 300-0370 300-0371 300-0368 300-0374 300-0376 300-0362 300-0366 300-0367 300-0363
Dosing spoons, white
Cucchiai
Volumetric and dosing spoons can be used to measure small amounts of powder, granulates and liquids exactly and dose them precisely. For precise measuring, excess medium is stripped off the upper edge of the spoon. The content volume is engraved on the handle.
  • PS, white
  • Optionally packed individually and sterilised
  • Stripper edge
  • For single use
  • Cleanroom manufactured (cleanroom Class 7)
The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom or getting into the corners of a container, thus allowing for efficient emptying of small remaining amounts. The flat base makes it possible to set the volumetric spoon down without spilling the contents.
The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities. Since this plastic volumetric spoon was developed for single use, there is no cleaning, and the possibility of cross-contamination and impurities is excluded.
These dosing and volumetric spoons are suitable for precise measurements in chemical and pharmaceutical applications, cosmetics, animal feed also the food and beverage industry.
Certificazioni: Complies with EU food requirements and FDA regulations.
Confezione: Available either in a large pack of 100 pieces or 100 pieces packed individually and radiation sterilised.