Stampa…
VWR® Economy Line 2.0, Laboratory Chairs
Fornitore: VWR Collection
132-1404EA 311 EUR
132-1404 132-1405 132-1406 132-1407 132-1408 132-1409
VWR® Economy Line 2.0, Laboratory Chairs
Sedie
Economy line 2.0 is a simple laboratory chair that still meets all requirements that a laboratory implies. The chair is ergonomic, robust, easy to clean, intuitive to use and has an excellent price-performance ratio.
  • A laboratory chair without any compromises
  • Specific laboratory ergonomics
  • Two upholstery options (artificial leather or PU foam): comfortable and easy to clean
  • Extremely robust
  • Unbeatable in price and performance
  • Optional 4D armrests
Reliable body support for laboratory work thanks to a permanent contact backrest. Seat height adjustment, backrest height adjustment by 60 mm, seat tilt adjustment by 6°, solid plastic star base, colour black.
Certificazioni: GS certified, suitable for use in biosafety laboratories BSL 1 and BSL 2, GMP conformity.
Informazioni per l'ordine: 10 year warranty. For optional armrests, contact your local Avantor representative.