Stampa…

Giemsa colorante soluzione, J.T.Baker®

Fornitore: Avantor
Pericolo

Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

3856.1000EA 45.3 EUR
3856.1000 3856.2500 3856.9180ST
Giemsa colorante soluzione, J.T.Baker®
Giemsa colorante

GIEMSA's solution Azur-Eosine-methylene blue solution for microscopy (contains methanol).

MDL Number: MFCD00081642
CAS Number: 51811-82-6
UN: 1992
ADR: 3,II
IVDLogo

Visualizza i codici

Risultati del test di specificazione

Performance Test Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR