Stampa…
Intelligent sole cleaning machine (ISCM)
Fornitore: GUANG DONG SUOREC TECHNOLOG
CLW-T10P CLW-T18P CLW-T28P CLW-T13P
113-9164EA 3710 EUR
113-9164 113-9166 113-9167 113-9165
Intelligent sole cleaning machine (ISCM)
Lucido da scarpe e spazzole
The Intelligent Sole Cleaning Machine (ISCM) effectively prevents embarrassing footprints by eliminating the need for sticky mat or shoe cover inventory, disposal, and replacement costs.
  • High particle removal efficiency up to 96%
  • Simple installation, and easy to clean
  • User-friendly and environmentally-friendly design
  • Long service life up to 8 years
  • Improve product yield and save cleanroom maintenance costs
Simple cleaning operation requires stepping onto the sole cleaning machine and remaining atop the cleaning grills for 3 seconds. Then step onto the drying mat and remain for 1 second.
High quality 316 stainless steel construction and a slim design ensure a long service life and allow use within an air shower. Water changing times can be arranged manually or by automation settings. Programmed water changing times can be set according to a programmed schedule or number of users. The touch panel controller features six different languages (Chinese, English, French, German, Spanish and Italian). An auto shutdown will occur in the event of electric leakage or over-maximum water levels. Triple-overflow protection includes photoelectric conductor, limit induction floating switch and program monitoring. The integrated power pump supports long distance drainage, can grind dirt, and will not be blocked when air is inhaled. Each brush belt includes eight stainless steel wire drawings and adopts a special stitching process, which will give belts a longer lifetime. All wiring is done by one single PCB. Pilot lights and a pilot light strips can also be installed.
Besides acting as a stand-alone device, the ISCM can be connected with an air shower, an ESD access control system and intelligent gate, and can be placed together for wheel carts. The ISCM is ideal for use in food manufacturing and automotive industries, as well as any workplace seeking to reduce disposable sticky mat use and improve environmental compliances. The ISCM will save transportation costs, waste disposable costs, warehouse costs, and cleaning time, all while improving product yield.
Certificazioni: CE certified.