Stampa…
N,O-Bis(trimetilsilil)acetammide silylation reagent, for GC derivatisation
Fornitore: MACHEREY-NAGEL
Pericolo
Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate
701210.110 701210.150 701210.510
MANA701210.110EA 39.8 EUR
MANA701210.110 MANA701210.150 MANA701210.510
N,O-Bis(trimetilsilil)acetammide silylation reagent, for GC derivatisation
N,O-Bis(trimetilsilil)acetammide
BSA is a strong silylation reagent that forms very stable TMS derivatives with a large variety of compounds, e.g., non-sterically hindered alcohols, carboxylic acids, phenols, enols, steroids, (biogenic) amines and alkaloids.
  • Silylation can be applied on many compounds
  • Silylation reagents are easily prepared
  • BSA derivatives have great volatility
Not recommended for use with carbohydrates or very low molecular weight compounds. Good solvent for polar compounds, but frequently used in combination with a solvent (pyridine, DMF etc.) or with other silylation reagents or catalysts such as TFA, HCl or TMBS. Used in combination with DMF, BSA is the reagent of choice for derivatizing phenols.
Formula: C₈H₂₁NOSi₂
Peso molecolare: 203.43 g/mol
Punto di ebollizione: 71–73 °C (35 mm Hg)
Punto di fusione: 24
Densità: 0.832 g/cm³ (d20°/4°)
Flash Pt: 40
MDL Number: MFCD00008270
CAS Number: 10416-59-8
UN: 3316
ADR: 9 II