Stampa…
1-(Heptafluorobutyryl)imidazole ≥98.5% (by GC), LiChropur™ for GC derivatisation, Supelco®
Fornitore: Merck
Synonyms: 1-(Perfluorobutanoyl)-1H-imidazole, N-Heptafluorobutyrylimidazole
74382-10X1ML 74382-10ML 74382-1ML
74382-10X1MLEA 485 EUR
74382-10X1ML 74382-10ML 74382-1ML
1-(Heptafluorobutyryl)imidazole ≥98.5% (by GC), LiChropur™ for GC derivatisation, Supelco®
1-(Heptafluorobutyryl)imidazole
Formula: C₇H₃F₇N₂O
Peso molecolare: 264,10 g/mol
MDL Number: MFCD00014503
CAS Number: 32477-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4ebc844bc9f067b7a2d38d7c3f7a0ccf4e24c27&vid=ecc81daad25f151efaabf82d5d16963bf912c46b&prid=web&clid=SAPDAM