Stampa…
Tubing, silicone
Fornitore: BURKLE
8803-0812 8803-0508 8803-0710 8803-0609
BURK8803-0812EA 55.8 EUR
BURK8803-0812 BURK8803-0508 BURK8803-0710 BURK8803-0609
Tubing, silicone
Tubazioni Tubazione di trasferimento
Silicone tubing is a versatile transparent hose.
  • Expands while using organic solvents
  • Autoclavable, ethylene oxide
  • Appropriate for food
  • Withstand temperature from −60 to +180 °C
  • Good resistance against weak acids
To the extend it is non-reactive to ionising solvents except chlorinated aliphatic and aromatic hydrocarbons.
Hardness (Shore A): 55 ±5
Attenzione: Should not use it for harsh solutions and strong acids.