Stampa…
Magnesium carbonate hydroxide pentahydrate leggero 98% (by complexometric analysis)
Fornitore: Thermo Scientific
Synonyms: Magnesium carbonate basic pentahydrate, Magnesium hydroxide carbonate pentahydrate
A18070.30 A18070.0B A18070.0I
A18070.30EA 24 EUR
A18070.30 A18070.0B A18070.0I
Magnesium carbonate hydroxide pentahydrate leggero 98% (by complexometric analysis)
Magnesium carbonate hydroxide pentahydrate
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.
Attenzione: Stable under recommended storage conditions. Incompatible with oxidsing agents.
Formula: C₄H₂Mg₅O₁₄·5H₂O
Peso molecolare: 485,65 g/mol (395,58 anhy)
Densità: 2,160 g/cm³
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149784
CAS Number: 56378-72-4
EINECS: 235-192-7
Visualizza i codici
Appearance (Colour) White
Form Powder
Complexometric Titration >97.5%