Stampa…
(±)11(12)-EET methyl ester soluzione ≥98% in ethanol
Fornitore: Cayman Chemical
Pericolo
Synonyms: (5Z,8Z,14Z)-11,12-Epoxy-5,8,14-icosatrienoic acid methyl ester, (+/-)11(12)-Epetre methyl ester
50510-100 50510-500 50510-25 50510-50
CAYM50510-100EA 511 EUR
CAYM50510-100 CAYM50510-500 CAYM50510-25 CAYM50510-50
(±)11(12)-EET methyl ester soluzione ≥98% in ethanol
(±)11(12)-EET methyl ester
(±)11(12)-EET is biosynthesized in rat and rabbit liver microsomes by CYP450. (±)11(12)-EET has been shown, along with (±)8(9)-EET, to play a role in the recovery of depleted Ca²⁺ pools in cultured smooth muscle cells.
(±)11(12)-EET methyl ester is a more stable for used for long-term storage. It can be readily hydrolysed to the free acid as needed.
Formula: C₂₁H₃₄O₃
Peso molecolare: 334,5 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00077342
CAS Number: 73799-06-1
UN: 1170
ADR: 3,II
Visualizza i codici
Purity ≥98%