Stampa…
Colorimetric rapid tests, silica, VISOCOLOR®
Fornitore: MACHEREY-NAGEL
920087 920187 931033 931233 931234
MANA920087EA 119 EUR
MANA920087 MANA920187 MANA931033 MANA931233 MANA931234
Colorimetric rapid tests, silica, VISOCOLOR®
Kit per analisi ambientali
The VISOCOLOR® testkits are devided into VISOCOLOR® alpha, ECO and HE. Every product range offering its specific strenght to provide a tailored test for each and every user and application.
  • Portable
  • Environmentally friendly
  • Convenient handling
  • Application-oriented measuring ranges
Natural water contains silica in different amounts, depending on the geological conditions the water encounters during its course. The silica occurs partly as soluble silicate, partly colloidal as polysilicic acids. The silica content in water for medium and high pressure boilers may not exceed certain limits; in power plants therefore constant measurement of the silica content is required.