Stampa…
Qualitative filter papers, grade 202, Whatman™
Fornitore: Whatman products (Cytiva)
5202-110 5202-090 5202-125 5202-150 5202-185 5202-240 5202-320 5202-400 5202-330 5202-250 5202-200
515-2829EA 11.3 EUR
515-2829 515-2828 515-2830 515-2831 515-2832 515-2833 515-2834 WHAT5202-400 WHAT5202-330 WHAT5202-250 WHAT5202-200
Qualitative filter papers, grade 202, Whatman™
Filtri Carta da filtro Carta da filtro per analisi qualitative
A general qualitative filter with creped surface. Faster than grade 201 and retains large crystalline coarse particles. Widely used in industrial laboratories and students laboratories for routine filtration.
  • Filters coarse particles
  • Medium filtration speed
  • Creped for increased efficiency
The filter is wet-strengthened with a small amount of chemically stable resin to extend the life of the filter and improve handling. Normal qualitative applications will not introduce any significant impurities into the filtrate. However it is not recommended for Kjeldahl nitrogen analysis.
Visualizza i codici
Specifiche
Grado 202
Environmentally Preferable
Peso 82 g/m²
Spessore 290 µm
Ritenzione delle particelle 15 - 19 µm