Stampa…
Chemical resistant gloves, Tychem® NT430
Fornitore: DuPont
NT4300BU080288 NT4300BU110288
112-5478EA 0 EUR
112-5478 112-5481
Chemical resistant gloves, Tychem® NT430
Guanti
These 100% nitrile gloves are the perfect hybrid for jobs requiring chemical performance plus optimum fit, feel and comfort.
  • Very comfortable hybrid nitrile
  • Lightweight with 'second skin' feel
  • Embossed pattern finish provides effective, long lasting grip
Ideal for working in damp or greasy environments and for small parts handling.
Certificazioni: EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016 (BACTERIA); Cat. III
Food contact approved.
Visualizza i codici
Specifiche
Modello NT430
Environmentally Preferable
Ambidestro/mano specifico mano specifico
Monouso/riutilizzabile monouso
Materiale rivestimento interno Unlined
Finitura Embossed
Tipo polsino Arrotolato
Norma EN EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016 (BACTERIA)
Approvati per il contatto con gli alimenti +