Stampa…
ESD safety shoes, lace-up, uvex 2, 6508
Fornitore: UVEX
6508.1/35 6508.1/36 6508.1/37 6508.1/38 6508.1/39 6508.1/40 6508.1/41 6508.1/42 6508.1/43 6508.1/44 6508.1/45 6508.1/46 6508.1/47 6508.1/48 6508.1/49 6508.1/50 6508.1/51 6508.1/52 6508.2/37 6508.2/38 6508.2/39 6508.2/40 6508.2/41 6508.2/42 6508.2/43 6508.2/44 6508.2/45 6508.2/35 6508.2/36 6508.2/46 6508.2/47 6508.2/48 6508.2/49 6508.2/50 6508.2/51 6508.2/52 6508.3/35 6508.3/36 6508.3/37 6508.3/38 6508.3/39 6508.3/40 6508.3/41 6508.3/42 6508.3/43 6508.3/44 6508.3/45 6508.3/46 6508.3/47 6508.3/48 6508.3/49 6508.3/50 6508.3/51 6508.3/52 6508.4/35 6508.4/36 6508.4/37 6508.4/38 6508.4/39 6508.4/40 6508.4/41 6508.4/42 6508.4/43 6508.4/44 6508.4/45 6508.4/46 6508.4/47 6508.4/48 6508.4/49 6508.4/50 6508.4/51 6508.4/52
UVEX6508.1/35EA 140 EUR
UVEX6508.1/35 UVEX6508.1/36 UVEX6508.1/37 UVEX6508.1/38 UVEX6508.1/39 UVEX6508.1/40 UVEX6508.1/41 UVEX6508.1/42 UVEX6508.1/43 UVEX6508.1/44 UVEX6508.1/45 UVEX6508.1/46 UVEX6508.1/47 UVEX6508.1/48 UVEX6508.1/49 UVEX6508.1/50 UVEX6508.1/51 UVEX6508.1/52 UVEX6508.2/37 UVEX6508.2/38 UVEX6508.2/39 UVEX6508.2/40 UVEX6508.2/41 UVEX6508.2/42 UVEX6508.2/43 UVEX6508.2/44 UVEX6508.2/45 UVEX6508.2/35 UVEX6508.2/36 UVEX6508.2/46 UVEX6508.2/47 UVEX6508.2/48 UVEX6508.2/49 UVEX6508.2/50 UVEX6508.2/51 UVEX6508.2/52 UVEX6508.3/35 UVEX6508.3/36 UVEX6508.3/37 UVEX6508.3/38 UVEX6508.3/39 UVEX6508.3/40 UVEX6508.3/41 UVEX6508.3/42 UVEX6508.3/43 UVEX6508.3/44 UVEX6508.3/45 UVEX6508.3/46 UVEX6508.3/47 UVEX6508.3/48 UVEX6508.3/49 UVEX6508.3/50 UVEX6508.3/51 UVEX6508.3/52 UVEX6508.4/35 UVEX6508.4/36 UVEX6508.4/37 UVEX6508.4/38 UVEX6508.4/39 UVEX6508.4/40 UVEX6508.4/41 UVEX6508.4/42 UVEX6508.4/43 UVEX6508.4/44 UVEX6508.4/45 UVEX6508.4/46 UVEX6508.4/47 UVEX6508.4/48 UVEX6508.4/49 UVEX6508.4/50 UVEX6508.4/51 UVEX6508.4/52
ESD safety shoes, lace-up, uvex 2, 6508
Scarpe e stivali
Metal-free, lightweight, flexible and robust safety shoe made from breathable, water-repellent high-tech microvelour upper material, soft padded tongue and collar, plus a dual-density PUR sole free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Virtually seam-free construction eliminates pressure points
  • Lightweight two-layer sole with excellent shock absorption and outstanding slip resistance
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Injection-moulded PU scuff cap for additional protection
  • Flexible, metal-free puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
For medium industrial applications including the chemical, transport, logistics, energy and mechanical engineering industries. Made from synthetic textiles so suitable for those allergic to chrome
Colour: Black/orange
Certificazioni: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S3, SRC
Informazioni per l'ordine: Normal laces also provided.