Stampa…
ESD safety ankle boots, lace-Up, Lex
Fornitore: ELTEN
766231/45 766231/38 766231/43 766231/40 766231/41 766231/47 766231/39 766231/44 766231/48 766231/46 766231/42
ELTN766231/45EA 83.9 EUR
ELTN766231/45 ELTN766231/38 ELTN766231/43 ELTN766231/40 ELTN766231/41 ELTN766231/47 ELTN766231/39 ELTN766231/44 ELTN766231/48 ELTN766231/46 ELTN766231/42
ESD safety ankle boots, lace-Up, Lex
Scarpe e stivali
ESD safety shoe made from cowhide, hydrophobised nubuck leather, a closed padded tongue with mesh inserts made of reflective materials and a dual-density PU/PU sole.
  • Climate regulating, breathable textile lining
  • Full length inlay sole (ESD Pro), black
  • Replaceable insole; orthopaedic insoles possible
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificazioni: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, Form B.