Menaquinone 7 ≥98%
Fornitore: Cayman Chemical
Synonyms:
Vitamin MK-7, Menaquinone-7, Vitamin K2(35), 2-((2E,6E,10E,14E,18E,22E)-3,7,11,15,19,23,27-Heptamethyloctacosa-2,6,10,14,18,22,26-heptaen-1-yl)-3-methylnaphthalene-1,4-dione, MK-7, 2-(-3,7,11,15,19,23,27-heptamethyloctacosa-2E,6E,10E,14E,18E,22E,26-heptaen-1-yl)-3-methylnaphthalene-1,4-dione
Menaquinone 7 (MK-7) is a vitamin K2 analog that has been found in a variety of fermented foods. It increases osteocalcin, osteoprotegerin, and RANKL mRNA expression in osteoblastic MC3T3E cells. MK-7 (1 µM) increases alkaline phosphatase activity and calcium content in femoral-metaphyseal tissues isolated from normal and skeletal-unloaded rats. In vivo, MK-7 (100 µg/g diet) inhibits aortic and myocardial calcification and decreases aortic alkaline phosphatase tissue concentrations in a rat model of extraosseous calcification. It also increases trabecular number, bone mineral density, and bone mineral content in a rat model of ovariectomy-induced bone loss.
- Formulation: A crystalline solid
- UV/Vis: λmax 248, 270, 330 nm
- Stability: ≥ 2 years
Attenzione: This product is not for human or veterinary use.
Formula:
C₄₆H₆₄O₂ Peso molecolare: 649,0 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD06200757 CAS Number: 2124-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...