Stampa…
Liquid chromatography sorbents, Aluminium Oxide 90
Fornitore: MACHEREY-NAGEL
815020.5 815030.1 815030.25 815030.5 815010.1 815010.25 815010.5 815020.1 815020.25
MANA815020.5EA 244 EUR
MANA815020.5 MANA815030.1 MANA815030.25 MANA815030.5 MANA815010.1 MANA815010.25 MANA815010.5 MANA815020.1 MANA815020.25
Liquid chromatography sorbents, Aluminium Oxide 90
Sorbenti per cromatografia Flash Sorbenti per HPLC
Aluminium oxide adsorbents for column chromatography.
  • High-quality
  • Activity grade I
  • Useful alternative to silica gels in liquid chromatography
Aluminium oxides produced by dehydration of different aluminium hydroxides, e.g., hydrargillite between 400 and 500 °C.
Specific surface (BET): 130 m²/g
Informazioni per l' accessorio: Accessories not included.
Visualizza i codici
Specifiche
Tipo Aluminium oxide 90 acidic Aluminium oxide 90 basic Aluminium oxide 90 neutral
Environmentally Preferable
pH di esercizio pH 4 ± 0,3 pH 9,5 ± 0,3 pH 7 ± 0,5