Stampa…
COD standard solutions, Tintometer®
2420804 2420803 2420805
LOVI2420804EA 43 EUR
LOVI2420804 LOVI2420803 LOVI2420805
COD standard solutions, Tintometer®
Standard Standard per analisi ambientali
Standard solutions are solutions with a defined concentration and are provided to check the operation methods and devices of the cuvette tests as well as the condition of optical filters and the instrument.