Stampa…
ULT chest freezers, CxF series, Revco®
Fornitore: REVCO TECHNOLOGIES
ULT1390-10D
REVCULT1390-10DEA 0 EUR
REVCULT1390-10D
ULT chest freezers, CxF series, Revco®
Congelatori Ultra freezer
Revco® ExF series ultra low temperature freezers are designed for daily sample protection and dependability.
  • Eye-level, centralized information center, including microprocessor control and monitoring system, ensures that all controls and displays are easy to reach and read
  • Inner sub-lids protect bulk samples from ambient exposure during sample retrievals
  • Heavy gauge, cold-rolled steel exterior construction with a powder coat paint finish that resists chipping and rust and 127 mm foamed-in-place,polyurethane insulation
  • Power Management System with low voltage surge protection and buck/boost
  • Access port: 2 (25 mm)
  • The chest freezers have heavy duty casters
Construction: Exterior: Steel, white coated and interior in Stainless steel.
Temperature range: −50 °C to −86°C
Informazioni per l'ordine: Optional CO₂ or LN₂ safety back-up system is available for additional protection in the event of a power or mechanical failure. Optional, built-in chart recorder or data logger for continuous monitoring can be ordered separately, for details of the range of accessories, please ontact your local Avantor sales office.