Stampa…
ESD safety ankle boots, lace-up, Maverick Black Mid
Fornitore: ELTEN
763391/38 763391/39 763391/40 763391/41 763391/42 763391/43 763391/44 763391/45 763391/46 763391/47 763391/48
114-8969EA 87.9 EUR
114-8969 114-8970 114-8971 114-8972 114-8973 114-8974 114-8975 114-8976 114-8977 114-8978 114-8979
ESD safety ankle boots, lace-up, Maverick Black Mid
Scarpe e stivali
Leather-free ESD safety ankle boots made from hydrophonised microfibre and hydrophobised textile material, skin friendly textile lining, a full padded bellows tongue and collar padding plus a dual-density PU/PU sole (Fun).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
  • Anatomically-shaped insole and ankle support for improved stability
  • Protective steel toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black or black/red
Certificazioni: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC