Stampa…
ESD safety shoes, sandals, Velcro® fastening, Scott XXT
Fornitore: ELTEN
719731/36 719731/37 719731/38 719731/39 719731/40 719731/41 719731/42 719731/43 719731/44 719731/45 719731/46 719731/47 719731/48
114-9059EA 95.9 EUR
114-9059 114-9060 114-9061 114-9062 114-9063 114-9064 114-9065 114-9066 114-9067 114-9068 114-9069 114-9070 114-9071
ESD safety shoes, sandals, Velcro® fastening, Scott XXT
Scarpe e stivali
100% metal- and leather-free, lightweight ESD safety sandal made from microfibre and textile upper, reflective materials, a padded tongue and a slip-resistant dual-density TPU/PU sole SPORTICS.
  • Climate regulating, breathable textile lining
  • Full-length inlay sole ESD PRO black
  • Soft fleece ESD insole
  • Non-metallic puncture resistant midsole
  • Composite toe protection cap (200 J)
  • Very wide version
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificazioni: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC