Stampa…
ESD safety shoes, lace-up, Senex S2
Fornitore: ELTEN
72872/35 72872/36 72872/37 72872/38 72872/39 72872/40 72872/41 72872/42 72872/43 72872/44 72872/45 72872/46 72872/47 72872/48
ELTN72872/35EA 81.9 EUR
ELTN72872/35 ELTN72872/36 ELTN72872/37 ELTN72872/38 ELTN72872/39 ELTN72872/40 ELTN72872/41 ELTN72872/42 ELTN72872/43 ELTN72872/44 ELTN72872/45 ELTN72872/46 ELTN72872/47 ELTN72872/48
ESD safety shoes, lace-up, Senex S2
Scarpe e stivali
100% metal- and leather-free, sporty, lightweight ESD safety shoe made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black
Certificazioni: EN 61340-5-1 (ESD);More Product Information