Stampa…
N-ɑ-Fmoc-N-Trityl-L-histidine, white to off-white powder
Fornitore: Novabiochem (Part of Merck)
Synonyms: N-ɑ-Fmoc-tele-trityl-L-histidine, Fmoc-His(Trt)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-trityl-1H-imidazol-4-yl)propanoic acid, N-ɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-tele-(triphenylmethyl)-L-histidine, N-Fmoc-1-trityl-L-histidine
8.52032.0100 8.52032.0025 8.52032.0500 8.52032.0250 8.52032.2000 8.52032.5000
04-12-1065-100EA 243 EUR
04-12-1065-100 04-12-1065-25 8.52032.0500 8.52032.0250 NOVA8.52032.2000 NOVA8.52032.5000
N-ɑ-Fmoc-N-Trityl-L-histidine, white to off-white powder
N-alpha-Fmoc-N-Trityl-L-histidine
  • Assay (acidimetric): ≥97,0%
  • Water (K.F.): ≤2,0%
Formula: C₄₀H₃₃N₃O₄
Peso molecolare: 619.72 g/mol
MDL Number: MFCD00043332
CAS Number: 109425-51-6