3-Bromo-2-hydroxypyridine ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₅H₄BrNO Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD03411569 CAS Number: 13466-43-8 |
|
Risultati del test di specificazione
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 178.0 to 186.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...