2-Amino-6-chloropurine 99%
Fornitore: Thermo Fisher Scientific
Synonyms:
6-Chloroguanine, 6-Chloro-7H-purin-2-ylamine
Purity: 99%
CAS No.: 10310-21-1
Molecular Formula: C5H4ClN5
Formula Weight: 169.57
Melting Point:
Formula:
C₅H₄ClN₅ Punto di fusione: >300 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00075252 CAS Number: 10310-21-1 EINECS: 233-686-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...