Stampa…

Ferro (II) bromuro, ultra dry ≥99.995% (metals basis)

Fornitore: Thermo Scientific
Pericolo

Synonyms: Iron dibromide, Ferrous bromide, Iron(II) bromide

Ampuled under argon

43472.14
43472.14EA 471 EUR
43472.14
Ferro (II) bromuro, ultra dry ≥99.995% (metals basis)
Ferro (II) bromuro

Purity: 99.995% CAS No.: 7789-46-0 Molecular Formula: Br2Fe Formula Weight: 215.67 Physical Form: Powder, Ampouled under argon Density: 4.636 Melting Point: 684° Merck Reference: 14,4042 UN No.: 3260 MDL No.: MFCD00016081

Formula: Br₂Fe
Peso molecolare: 215,66 g/mol
Punto di fusione: 684 °C
Densità: 4,636
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016081
CAS Number: 7789-46-0
EINECS: 232-168-8
UN: 3260
ADR: 8,III
Merck Index: 14,4042

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR