Stampa…
Cobalt(II) bromide, ultra dry ≥99.99% (metals basis)
Fornitore: Thermo Fisher Scientific
Pericolo
Synonyms: Cobaltous bromide
Ampuled under argon
40319.03 40319.06
40319.03EA 138 EUR
40319.03 40319.06
Cobalt(II) bromide, ultra dry ≥99.99% (metals basis)
Cobalt(II) bromide
Purity: 99.99% CAS No.: 7789-43-7 Molecular Formula: CoBr2 Formula Weight: 218.75 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 4.909 Appearance: Green Melting Point: 678° (in N2) Merck Reference: 14,2435 MDL No.: MFCD00016017
Formula: Br₂Co
Peso molecolare: 218.74 g/mol
Punto di fusione: 678 °C (in N{2})
Densità: 4.909
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016017
CAS Number: 7789-43-7
EINECS: 232-166-7
Merck Index: 14,2435