Stampa…

Gold(I) iodide ≥99%

Fornitore: Thermo Fisher Scientific
Pericolo

16617.03 16617.06
16617.03EA 204 EUR
16617.03 16617.06
Gold(I) iodide ≥99%
Gold(I) iodide

Purity: 99% CAS No.: 10294-31-2 Molecular Formula: AuI Formula Weight: 323.87 Physical Form: Crystalline Density: 8.25 Melting Point: 120° dec. Solubility: Insoluble in water. Soluble in alkali iodide, cyanide solutions. Decomposes in warm acids UN No.: 3260 MDL No.: MFCD00014175

Formula: AuI
Punto di fusione: 120 °C (dec.)
Densità: 8,25
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00014175
CAS Number: 10294-31-2
EINECS: 233-656-3
UN: 3260
ADR: 8,III
Merck Index: 12,04530

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR