Rame (I) bromuro ≥99.998% (metals basis), Puratronic®
Fornitore: Thermo Scientific
|
Attenzione
|
Ampuled under argon
Purity: 99.998%
CAS No.: 7787-70-4
Molecular Formula: CuBr
Formula Weight: 143.45
Physical Form: Powder, Ampouled under argon
Density: 4.98
Appearance: Pale green
Melting Point: 492°
Boiling Point: 1345°
Merck Reference: 14,2659
UN No.: 3077
MDL No.: MFCD00010969
Formula:
BrCu Peso molecolare: 143,45 g/mol Punto di ebollizione: 1345 °C Punto di fusione: 492 °C Densità: 4,98 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00010969 CAS Number: 7787-70-4 EINECS: 232-131-6 UN: 3077 ADR: 9,III Merck Index: 14,2659 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...