Stampa…

Tulio (III) ossido ≥99.9% (REO, rare earth oxide basis), REacton®

Fornitore: Thermo Fisher Scientific
Attenzione

11198.04 11198.09 11198.18
11198.04EA 152 EUR
11198.04 11198.09 11198.18
Tulio (III) ossido ≥99.9% (REO, rare earth oxide basis), REacton®
Tulio (III) ossido

Purity: 99.9% CAS No.: 12036-44-1 Molecular Formula: O3Tm2 Formula Weight: 385.87 Physical Form: Powder Appearance: Pale green Melting Point: 2425° Solubility: Dissolves slowly in strong acids Merck Reference: 14,9394 MDL No.: MFCD00011285

Formula: O₃Tm₂
Peso molecolare: 385.87 g/mol
Punto di fusione: 2425 °C
Densità: 8.6 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011285
CAS Number: 12036-44-1
EINECS: 234-851-6
Merck Index: 14,9394

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR