Stampa…
Terbium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Fornitore: Thermo Fisher Scientific
Attenzione
Ampuled under argon
35798.03 35798.09
35798.03EA 103 EUR
35798.03 35798.09
Terbium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Terbium(III) chloride
Purity: 99.99% CAS No.: 10042-88-3 Molecular Formula: TbCl3 Formula Weight: 265.29 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic Merck Reference: 14,9157 MDL No.: MFCD00011258
Formula: Cl₃Tb
Peso molecolare: 265.28 g/mol
Punto di fusione: 588
Densità: 4.35 g/cm³ (25 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011258
CAS Number: 10042-88-3
EINECS: 233-132-4
Merck Index: 14,9157