Indole-2-carboxylic acid 99%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Formula:
C₉H₇NO₂ Punto di fusione: 205…209 °C |
MDL Number:
MFCD00005611 CAS Number: 1477-50-5 EINECS: 216-030-4 |
Risultati del test di specificazione
Appearance | Off-white to yellow to brown Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 205 to 209 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...