Stampa…

Ethyl methyl sulphide ≥97%

Fornitore: Thermo Fisher Scientific
Pericolo

L02859.14 L02859.22
L02859.14EA 58.4 EUR
L02859.14 L02859.22
Ethyl methyl sulphide ≥97%
Ethyl methyl sulphide

Purity: 97% CAS No.: 624-89-5 Molecular Formula: C3H8S Formula Weight: 76.16 Flash Point: -15°(5°F) Refractive Index: 1.4392 Odour: Stench Melting Point: -106° Boiling Point: 66-67° Beilstein Reference: 1696871 UN No.: 1993 MDL No.: MFCD00009268

Formula: C₃H₈S
Punto di ebollizione: 66…67 °C
Punto di fusione: –106 °C
Densità: 0,841
Flash Pt: –15 °C (5 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00009268
CAS Number: 624-89-5
EINECS: 210-868-4
UN: 1993
ADR: 3,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR