di-Ammonio tiosolfato 96%
Fornitore: Thermo Fisher Scientific
Attenzione
|
Synonyms:
Ammonium hyposulphite, Ammonium thiosulphate
Purity: 98+%
CAS No.: 7783-18-8
Molecular Formula: (NH4)2S2O3
Formula Weight: 148.20
pH: 6.0-6.5
Melting Point: ca 180° dec.
MDL No.: MFCD00015965
Formula:
H₈N₂O₃S₂ Punto di fusione: ∼ 180 °C (dec.) Densità: 1,64 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00015965 CAS Number: 7783-18-8 EINECS: 231-982-0 Merck Index: 14,562 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...