Stampa…

Chlorodiphenylphosphine 98+%, in astine

Fornitore: Thermo Scientific
Pericolo

Synonyms: DPPC, Diphenylchlorophosphine

41352.09 41352.18 41352.30
41352.09EA 33.9 EUR
41352.09 41352.18 41352.30
Chlorodiphenylphosphine 98+%, in astine
Chlorodiphenylphosphine

Purity: 98+% CAS No.: 1079-66-9 Molecular Formula: C12H10ClP Formula Weight: 220.64 Flash Point: 138°(280°F) Physical Form: Liquid Density: 1.204 Refractive Index: 1.6360 Sensitivity: Air and Moisture Sensitive Appearance: Colourless to pale yellow Odour: Acrid Melting Point: 14-16° Boiling Point: 318-321° Beilstein Reference: 512032 UN No.: 3265 MDL No.: MFCD00000529

Formula: C₁₂H₁₀ClP
Peso molecolare: 220,64 g/mol
Punto di ebollizione: 318…321 °C
Punto di fusione: 14…16 °C
Densità: 1,204
Flash Pt: 138 °C (280 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00000529
CAS Number: 1079-66-9
EINECS: 214-093-2
UN: 3265
ADR: 8,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR