Tricyclohexylphosphine ≥96%
Fornitore: Thermo Scientific
|
Attenzione
|
30386.06
30386.14
30386.03
30386.06EA
107
EUR
30386.06
30386.14
30386.03
Tricyclohexylphosphine ≥96%
Tricyclohexylphosphine
Purity: 96%
CAS No.: 2622-14-2
Molecular Formula: (C6H11)3P
Formula Weight: 280.42
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: White to off-white
Odour: Unpleasant
Melting Point: 76-78°
MDL No.: MFCD00003853
Formula:
C₁₈H₃₃P Peso molecolare: 280,43 g/mol Punto di ebollizione: 132…134 °C (0,02 mmHg) Punto di fusione: 76…78 °C Densità: 0,9 g/cm³ (20 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00003853 CAS Number: 2622-14-2 EINECS: 220-069-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...