Stampa…

Tetrabutylammonium nitrate ≥98%

Fornitore: Thermo Scientific
Pericolo

43342.14 43342.06 43342.22 43342.36
43342.14EA 143 EUR
43342.14 43342.06 43342.22 43342.36
Tetrabutylammonium nitrate ≥98%
Tetrabutylammonium nitrate

Purity: 98% CAS No.: 1941-27-1 Molecular Formula: [CH3(CH2)3]4NNO3 Formula Weight: 304.48 Physical Form: Powder Sensitivity: Hygroscopic UN No.: 1479 MDL No.: MFCD00043202

Formula: C₁₆H₃₆N₂O₃
Peso molecolare: 304,47 g/mol
Punto di fusione: 116…118 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00043202
CAS Number: 1941-27-1
EINECS: 217-726-0
UN: 1479
ADR: 5.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR