Trihexyl phosphate ≥90%
Fornitore: Thermo Scientific
|
Attenzione
|
16801.06
16801.14
16801.03
16801.06EA
109
EUR
16801.06
16801.14
16801.03
Trihexyl phosphate ≥90%
Trihexyl phosphate
Purity: 90+%
CAS No.: 2528-39-4
Molecular Formula: C18H39O4P
Formula Weight: 350.48
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00015351
Formula:
C₁₈H₃₉O₄P Peso molecolare: 350,48 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00015351 CAS Number: 2528-39-4 EINECS: 219-774-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...