Stampa…

Butyltin trichloride ≥96%

Fornitore: Thermo Scientific
Pericolo

Synonyms: n-Butyltin trichloride, Trichlorobutylstannane

71125.14 71125.22
71125.14EA 37.8 EUR
71125.14 71125.22
Butyltin trichloride ≥96%
Butyltin trichloride

Purity: 96% CAS No.: 1118-46-3 Molecular Formula: CH3(CH2)3SnCl3 Formula Weight: 282.17 Flash Point: 81°(178°F) Physical Form: Liquid Density: 1.693 Refractive Index: 1.5244 Sensitivity: Moisture Sensitive Odour: Acrid Melting Point: -63° Boiling Point: 98°/10mm UN No.: 3265 MDL No.: MFCD00000515

Formula: C₄H₉Cl₃Sn
Peso molecolare: 282,18 g/mol
Punto di ebollizione: 98 °C (10 mmHg)
Punto di fusione: –63 °C
Densità: 1,693
Flash Pt: 81 °C (178 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00000515
CAS Number: 1118-46-3
EINECS: 214-263-6
UN: 3265
ADR: 8,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR