Stampa…
Silicon(IV) oxide, amorphous S.A. 105-145 m²/g, fumed, surface treated
Fornitore: Thermo Scientific
Attenzione
Synonyms: Silica gel amorphous, Silicon dioxide amorphous, Silicium dioxide amorphous
42783.18 42783.30 42783.A1
42783.18EA 21 EUR
42783.18 42783.30 42783.A1
Silicon(IV) oxide, amorphous S.A. 105-145 m²/g, fumed, surface treated
Silicon(IV) oxide, amorphous
CAS No.: 7631-86-9 Molecular Formula: O2Si Formula Weight: 60.09 Physical Form: -325 mesh powder Density: 2.2 Appearance: White Odour: Odourless MDL No.: MFCD00011232
Adhesives, coatings, sealants, inks, thermal insulation, silicones to provide hydrophobicity
Formula: SiO₂
Peso molecolare: 60,08 g/mol
Punto di fusione: ~ 2230 °C
Densità: 2,2
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4