Alluminio ossido, aerosol refractory brushable paint
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
Aluminium(III) oxide, Alumina C
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Flash Point: -4°(25°F)
Physical Form: Paste
Appearance: White
UN No.: 1263
MDL No.: MFCD00003424
Provides protective high temperature Al2O3 coating on ceramic, graphite, or metal
Formula:
Al₂O₃ Peso molecolare: 101,96 g/mol Punto di ebollizione: 2980 °C (1013 hPa) Punto di fusione: 2045 °C Densità: 3,94 g/cm³ (20 °C) Flash Pt: –4 °C (25 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 UN: 1263 ADR: 3,II REACH: 01-2119529248-35 Merck Index: 13,00355 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...