Stampa…

Aluminium fluoride, anidro ≥99.99% (metals basis)

Fornitore: Thermo Scientific
Pericolo

Synonyms: Aluminum fluoride

44435.09 44435.18 44435.30
44435.09EA 116 EUR
44435.09 44435.18 44435.30
Aluminium fluoride, anidro ≥99.99% (metals basis)
Aluminium fluoride

Purity: 99.99% CAS No.: 7784-18-1 Molecular Formula: AlF3 Formula Weight: 83.98 Physical Form: Powder Density: 2.882 Melting Point: 1291° subl. Merck Reference: 14,339 MDL No.: MFCD00003426

Formula: AlF₃
Peso molecolare: 83,98 g/mol
Punto di fusione: 1291 °C (subl.)
Densità: 2,882
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003426
CAS Number: 7784-18-1
EINECS: 232-051-1
UN: 3260
ADR: 8,II
Merck Index: 14,339

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR