Aluminium bromide ≥99.997% (metals basis)
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Synonyms:
Aluminum bromide
Purity: 99.997%
CAS No.: 7727-15-3
Molecular Formula: AlBr3
Formula Weight: 266.71
Physical Form: Crystalline
Density: 3.21
Appearance: White to orange
Odour: Pungent
Melting Point: 97.5°
Boiling Point: 265°
Merck Reference: 14,332
UN No.: 1725
MDL No.: MFCD00003421
Formula:
AlBr₃ Peso molecolare: 266.69 g/mol Punto di ebollizione: 265 °C Punto di fusione: 97.5 °C Densità: 3.21 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00003421 CAS Number: 7727-15-3 EINECS: 231-779-7 UN: 1725 ADR: 8 II Merck Index: 14,332 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...