Stampa…

Sodium selenide ≥99.8% (metals basis)

Fornitore: Thermo Fisher Scientific
Pericolo

36187.03 36187.06
36187.03EA 374 EUR
36187.03 36187.06
Sodium selenide ≥99.8% (metals basis)
Sodium selenide

Purity: 99.8% CAS No.: 1313-85-5 Molecular Formula: Na2Se Formula Weight: 124.94 Physical Form: -60 Mesh (dry under argon) Density: 2.625 Appearance: Grey to pink Melting Point: 875° Merck Reference: 14,8672 UN No.: 3283 MDL No.: MFCD00014240

Formula: Na₂Se
Punto di fusione: >875 °C
Densità: 2,625
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00014240
CAS Number: 1313-85-5
EINECS: 215-212-0
UN: 3283
ADR: 6.1,II
Merck Index: 14,8672

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR