Stampa…
, anidro ≥98%, white powder
Fornitore: MP Biomedicals
Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
0215194505 02151945.1 02151945.5 0215194501
ICNA0215194505EA 500 EUR
ICNA0215194505 ICNA02151945.1 ICNA02151945.5 ICNA0215194501
, anidro ≥98%, white powder
di-Potassio idrogenofosfato
Soluble in water (1g/20 ml); Insoluble to slightly soluble in ethanol; insoluble in alcohol.
Formula: K₂HPO₄
Peso molecolare: 174.18 g/mol
Punto di fusione: 340
Densità: 2.3 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM