Cyclopentylmethanol ≥98%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
Cyclopentanemethanol
Purity: 98%
CAS No.: 3637-61-4
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 62°(143°F)
Refractive Index: 1.4580
Boiling Point: 162-163°
Beilstein Reference: 1919000
MDL No.: MFCD00001384
Formula:
C₆H₁₂O Peso molecolare: 100,16 g/mol Punto di ebollizione: 162…163 °C Densità: 0,925 Flash Pt: 62 °C (143 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00001384 CAS Number: 3637-61-4 EINECS: 222-861-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...