3-Methylbenzylamine 98%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
m-Tolylmethanamine, 3-(Aminomethyl)toluene
Purity: 98%
CAS No.: 100-81-2
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 80°(176°F)
Refractive Index: 1.5360
Boiling Point: 203-205°
Beilstein Reference: 507474
UN No.: 2735
MDL No.: MFCD00008118
Formula:
C₈H₁₁N Peso molecolare: 121,18 g/mol Punto di ebollizione: 203…205 °C Densità: 0,960 Flash Pt: 80 °C (176 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008118 CAS Number: 100-81-2 EINECS: 202-890-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...