4-Benzoylbenzoic acid 99%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
105720050
105720250
ACRO105720050EA
66.4
EUR
ACRO105720050
ACRO105720250
4-Benzoylbenzoic acid 99%
4-Benzoylbenzoic acid
Formula:
C₁₄H₁₀O₃ Punto di ebollizione: 416 °C (1013 hPa) Punto di fusione: 197…200 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00002560 CAS Number: 611-95-0 EINECS: 210-286-0 |
Risultati del test di specificazione
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 197 to 200 °C |
Titration with NaOH | ≥98.5 % |
UV | at 375 nm A: ≤0.15 |
Viscosity | 0.22 to 0.27 (1 % at 37 °C) |
Ash | ≤2.0 % |
Average molecular weight | 60000 to 90000 |
pH | 4.5 to 7.0 (6 % in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...