2-Benzoylbenzoic acid 98+%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Formula:
C₁₄H₁₀O₃ Punto di ebollizione: 436 °C (1013 hPa) Punto di fusione: 126…130 °C |
MDL Number:
MFCD00002472 CAS Number: 85-52-9 EINECS: 201-612-2 |
Risultati del test di specificazione
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 126 to 130 °C |
Titration with NaOH | ≥98.0 % |
UV | at 375 nm A: ≤0.15 |
Viscosity | 0.4 to 0.5 (1 % at 37 °C) |
Average molecular weight | 250000 |
pH | 4.5 to 7.0 (6 % in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...