Stampa…

3-Nitrobenzyl alcohol ≥99%

Fornitore: Thermo Scientific

Synonyms: (3-Nitrophenyl)methanol

44445.03 44445.06
44445.03EA 36.7 EUR
44445.03 44445.06
3-Nitrobenzyl alcohol ≥99%
3-Nitrobenzyl alcohol

Purity: 99+% CAS No.: 619-25-0 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 110°(230°F) Physical Form: Solid Density: 1.300 Refractive Index: 1.5730 Appearance: Yellow Melting Point: 29-33° Boiling Point: 175-180°/3mm Beilstein Reference: 2044769 MDL No.: MFCD00007273

Formula: C₇H₇NO₃
Peso molecolare: 153,14 g/mol
Punto di ebollizione: 175…180 °C (3 mmHg)
Punto di fusione: 29…33 °C
Densità: 1,300
Flash Pt: >110 °C (230 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007273
CAS Number: 619-25-0
EINECS: 210-588-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR