Stampa…

α,α,α-Trifluoro-o-cresol ≥98%

Fornitore: Thermo Scientific
Pericolo

Synonyms: 2-Hydroxybenzotrifluoride, 2-(Trifluoromethyl)phenol

A14231.03 A14231.06 A14231.14
A14231.03EA 46.4 EUR
A14231.03 A14231.06 A14231.14
α,α,α-Trifluoro-o-cresol ≥98%
α,α,α-Trifluoro-o-cresol

Purity: 98% CAS No.: 444-30-4 Molecular Formula: C7H5F3O Formula Weight: 162.11 Flash Point: 65°(149°F) Odour: Phenol-like Boiling Point: 147-148° Beilstein Reference: 1867917 UN No.: 3261 MDL No.: MFCD00002222

Formula: C₇H₅F₃O
Peso molecolare: 162,11 g/mol
Punto di ebollizione: 147…148 °C
Punto di fusione: 45…46 °C
Densità: >1 g/cm³ (20 °C)
Flash Pt: 65 °C (149 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002222
CAS Number: 444-30-4
EINECS: 207-148-7
UN: 3261
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR